4'-Cyanbiphenyl-4-carbonsäure

Lieferant: FLUOROCHEM

Synonyme: 4'-Cyano[1,1'-biphenyl]-4-carboxylic acid, 4'-Cyano-[1,1'-biphenyl]-4-carboxylic acid

020754-1G
FLUO020754-1GEA 956 EUR
FLUO020754-1G
4'-Cyanbiphenyl-4-carbonsäure
4'-Cyanbiphenyl-4-carbonsäure
Formel: C₁₄H₉NO₂
Molecular Weight: 223.23 g/mol
MDL: MFCD00438676
CAS-Nummer: 5728-46-1

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR