4'-Cyanbiphenyl-3-carbonsäure
Lieferant: FLUOROCHEM
020755-1G
FLUO020755-1GEA
143
EUR
FLUO020755-1G
4'-Cyanbiphenyl-3-carbonsäure
4'-Cyanbiphenyl-3-carbonsäure
Formel:
C₁₄H₉NO₂ |
MDL:
MFCD04117352 CAS-Nummer: 149506-93-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...