4'-Cyanbiphenyl-2-carbonsäure

Lieferant: FLUOROCHEM

020760-1G
FLUO020760-1GEA 590 EUR
FLUO020760-1G
4'-Cyanbiphenyl-2-carbonsäure
4'-Cyanbiphenyl-2-carbonsäure
Formel: C₁₄H₉NO₂
MDL: MFCD04117356
CAS-Nummer: 67856-50-2

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR