3'-Cyanbiphenyl-4-carbonsäure
Lieferant: FLUOROCHEM
020763-1G
FLUO020763-1GEA
527
EUR
FLUO020763-1G
3'-Cyanbiphenyl-4-carbonsäure
3'-Cyanbiphenyl-4-carbonsäure
Formel:
C₁₄H₉NO₂ |
CAS-Nummer:
5728-45-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...