3'-Cyanbiphenyl-3-carbonsäure
Lieferant: FLUOROCHEM
020764-1G
FLUO020764-1GEA
483
EUR
FLUO020764-1G
3'-Cyanbiphenyl-3-carbonsäure
3'-Cyanbiphenyl-3-carbonsäure
Formel:
C₁₄H₉NO₂ |
MDL:
MFCD04117360 CAS-Nummer: 253878-93-2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...