3'-Cyanbiphenyl-2-carbonsäure
Lieferant: FLUOROCHEM
020771-1G
FLUO020771-1GEA
1130
EUR
FLUO020771-1G
3'-Cyanbiphenyl-2-carbonsäure
3'-Cyanbiphenyl-2-carbonsäure
Formel:
C₁₄H₉NO₂ |
MDL:
MFCD04117367 CAS-Nummer: 107916-96-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...