4'-Hydroxybiphenyl-4-carbonsäure

Lieferant: FLUOROCHEM

Synonyme: 4'-Hydroxy-4-biphenylcarboxylic acid, 4'-Hydroxy-[1,1'-biphenyl]-4-carboxylic acid

020772-1G
FLUO020772-1GEA 51.8 EUR
FLUO020772-1G
4'-Hydroxybiphenyl-4-carbonsäure
4'-Hydroxybiphenyl-4-carbonsäure
Formel: C₁₃H₁₀O₃
Molecular Weight: 214,22 g/mol
MDL: MFCD00059078
CAS-Nummer: 58574-03-1

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR