4'-Hydroxybiphenyl-3-carbonsäure

Lieferant: FLUOROCHEM

020773-1G
FLUO020773-1GEA 536 EUR
FLUO020773-1G
4'-Hydroxybiphenyl-3-carbonsäure
4'-Hydroxybiphenyl-3-carbonsäure
Formel: C₁₃H₁₀O₃
MDL: MFCD01910126
CAS-Nummer: 121629-21-8

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR