4'-Hydroxybiphenyl-2-carbonsäure

Lieferant: FLUOROCHEM

Synonyme: 4'-Hydroxy-[1,1'-biphenyl]-2-carboxylic acid

020774-1G
FLUO020774-1GEA 411 EUR
FLUO020774-1G
4'-Hydroxybiphenyl-2-carbonsäure
4'-Hydroxybiphenyl-2-carbonsäure
Formel: C₁₃H₁₀O₃
Molecular Weight: 214,22 g/mol
MDL: MFCD04117368
CAS-Nummer: 67526-82-3

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR