3'-Hydroxybiphenyl-4-carbonsäure

Lieferant: FLUOROCHEM

020775-1G
FLUO020775-1GEA 288 EUR
FLUO020775-1G
3'-Hydroxybiphenyl-4-carbonsäure
3'-Hydroxybiphenyl-4-carbonsäure
Formel: C₁₃H₁₀O₃
CAS-Nummer: 220950-35-6

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR