3'-Hydroxybiphenyl-3-carbonsäure

Lieferant: FLUOROCHEM

Synonyme: 3'-Hydroxy-[1,1'-biphenyl]-3-carboxylic acid

020776-1G
FLUO020776-1GEA 438 EUR
FLUO020776-1G
3'-Hydroxybiphenyl-3-carbonsäure
3'-Hydroxybiphenyl-3-carbonsäure
Formel: C₁₃H₁₀O₃
Molecular Weight: 214,22 g/mol
MDL: MFCD04117370
CAS-Nummer: 171047-01-1

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR