3'-Hydroxybiphenyl-2-carbonsäure

Lieferant: FLUOROCHEM

Synonyme: 3'-Hydroxy-[1,1'-biphenyl]-2-carboxylic acid

020777-1G
FLUO020777-1GEA 252 EUR
FLUO020777-1G
3'-Hydroxybiphenyl-2-carbonsäure
3'-Hydroxybiphenyl-2-carbonsäure
Formel: C₁₃H₁₀O₃
Molecular Weight: 214,22 g/mol
MDL: MFCD04117371
CAS-Nummer: 92379-10-7

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR