3'-Acetylbiphenyl-4-carbonsäure

Lieferant: FLUOROCHEM

020790-1G
FLUO020790-1GEA 374 EUR
FLUO020790-1G
3'-Acetylbiphenyl-4-carbonsäure
3'-Acetylbiphenyl-4-carbonsäure
Formel: C₁₅H₁₂O₃
MDL: MFCD03839959
CAS-Nummer: 199678-04-1

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR