3'-Acetylbiphenyl-3-carbonsäure

Lieferant: FLUOROCHEM

020791-1G
FLUO020791-1GEA 588 EUR
FLUO020791-1G
3'-Acetylbiphenyl-3-carbonsäure
3'-Acetylbiphenyl-3-carbonsäure
Formel: C₁₅H₁₂O₃
MDL: MFCD03839958
CAS-Nummer: 728918-66-9

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR