3'-Acetylbiphenyl-2-carbonsäure
Lieferant: FLUOROCHEM
020792-1G
FLUO020792-1GEA
398
EUR
FLUO020792-1G
3'-Acetylbiphenyl-2-carbonsäure
3'-Acetylbiphenyl-2-carbonsäure
Formel:
C₁₅H₁₂O₃ |
MDL:
MFCD03839957 CAS-Nummer: 408367-33-9 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...