2',4'-Difluorbiphenyl-4-carbonsäure
Lieferant: FLUOROCHEM
020798-1G
FLUO020798-1GEA
248
EUR
FLUO020798-1G
2',4'-Difluorbiphenyl-4-carbonsäure
2',4'-Difluorbiphenyl-4-carbonsäure
Formel:
C₁₃H₈F₂O₂ Molecular Weight: 234,2 g/mol |
CAS-Nummer:
331760-41-9 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...