2',4'-Difluorbiphenyl-3-carbonsäure

Lieferant: FLUOROCHEM

020799-1G
FLUO020799-1GEA 345 EUR
FLUO020799-1G
2',4'-Difluorbiphenyl-3-carbonsäure
2',4'-Difluorbiphenyl-3-carbonsäure
Formel: C₁₃H₈F₂O₂
CAS-Nummer: 656304-77-7

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR