3'-Formylbiphenyl-3-carbonsäuremethylester

Lieferant: FLUOROCHEM

020807-1G
FLUO020807-1GEA 971 EUR
FLUO020807-1G
3'-Formylbiphenyl-3-carbonsäuremethylester
3'-Formylbiphenyl-3-carbonsäuremethylester
Formel: C₁₇H₁₆O₃
MDL: MFCD04117395
CAS-Nummer: 728918-83-0

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR