Methyl 4-(4-formylphenyl)benzoate
Lieferant: FLUOROCHEM
Synonyme:
4'-Formylbiphenyl-4-carbonsäuremethylester
020821-1G
FLUO020821-1GEA
0
EUR
FLUO020821-1G
Methyl 4-(4-formylphenyl)benzoate
Methyl 4-(4-formylphenyl)benzoate
Formel:
C₁₅H₁₂O₃ Molecular Weight: 240,25 g/mol |
MDL:
MFCD04039123 CAS-Nummer: 70916-89-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...