4'-(Trifluormethoxy)biphenyl-3-carbonsäure
Lieferant: FLUOROCHEM
020828-1G
FLUO020828-1GEA
321
EUR
FLUO020828-1G
4'-(Trifluormethoxy)biphenyl-3-carbonsäure
4'-(Trifluormethoxy)biphenyl-3-carbonsäure
Formel:
C₁₄H₉F₃O₃ Molecular Weight: 282,22 g/mol |
MDL:
MFCD04117416 CAS-Nummer: 728919-12-8 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...