2',4'-Dimethoxybiphenyl-4-carbonsäure
Lieferant: FLUOROCHEM
020836-1G
FLUO020836-1GEA
681
EUR
FLUO020836-1G
2',4'-Dimethoxybiphenyl-4-carbonsäure
2',4'-Dimethoxybiphenyl-4-carbonsäure
Formel:
C₁₅H₁₄O₄ |
CAS-Nummer:
108474-22-2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...