2',4'-Dimethoxybiphenyl-3-carbonsäure

Lieferant: FLUOROCHEM

020837-1G
FLUO020837-1GEA 1130 EUR
FLUO020837-1G
2',4'-Dimethoxybiphenyl-3-carbonsäure
2',4'-Dimethoxybiphenyl-3-carbonsäure
Formel: C₁₅H₁₄O₄
MDL: MFCD04117424
CAS-Nummer: 728919-16-2

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR