3',4'-Dimethylbiphenyl-4-carbonsäure
Lieferant: FLUOROCHEM
020842-1G
FLUO020842-1GEA
1130
EUR
FLUO020842-1G
3',4'-Dimethylbiphenyl-4-carbonsäure
3',4'-Dimethylbiphenyl-4-carbonsäure
Formel:
C₁₅H₁₄O₂ Molecular Weight: 226.27 g/mol |
CAS-Nummer:
122294-09-1 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...