3',4'-Dimethylbiphenyl-3-carbonsäure
Lieferant: FLUOROCHEM
020843-1G
FLUO020843-1GEA
1130
EUR
FLUO020843-1G
3',4'-Dimethylbiphenyl-3-carbonsäure
3',4'-Dimethylbiphenyl-3-carbonsäure
Formel:
C₁₅H₁₄O₂ |
MDL:
MFCD04117430 CAS-Nummer: 728919-21-9 |
Learn more
![](/m-de.vwr.com/de_DE/images/about_vwr_275x315.jpg)
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...