(3-Chlorphenyl)methansulfonsäurechlorid

Lieferant: FLUOROCHEM

Synonyme: 3-Chlorobenzylsulphonyl chloride, 3-Chloro-α-toluenesulphonyl chloride

020875-1G
FLUO020875-1GEA 60.9 EUR
FLUO020875-1G
(3-Chlorphenyl)methansulfonsäurechlorid
(3-Chlorophenyl)methanesulphonyl chloride
Formel: C₇H₆Cl₂O₂S
Molecular Weight: 225,09 g/mol
MDL: MFCD02683111
CAS-Nummer: 24974-73-0

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR