(2-Fluorphenyl)methansulfonsäurechlorid
Lieferant: FLUOROCHEM
020881-1G
FLUO020881-1GEA
119
EUR
FLUO020881-1G
(2-Fluorphenyl)methansulfonsäurechlorid
(2-Fluorophenyl)methanesulphonyl chloride
Formel:
C₇H₆ClFO₂S |
MDL:
MFCD04117455 CAS-Nummer: 24974-71-8 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...