(2-Methylphenyl)methansulfonsäurechlorid

Lieferant: FLUOROCHEM

Synonyme: 2-Methylbenzylsulphonyl chloride, o-Tolylmethanesulphonyl chloride

020884-1G
FLUO020884-1GEA 389 EUR
FLUO020884-1G
(2-Methylphenyl)methansulfonsäurechlorid
(2-Methylphenyl)methanesulphonyl chloride
Formel: C₈H₉ClO₂S
Molecular Weight: 204.673 g/mol
MDL: MFCD02683108
CAS-Nummer: 92614-55-6

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR