4-Fluorphenethylbromid

Lieferant: FLUOROCHEM

Synonyme: 1-Fluoro-4-(2-bromoethyl)benzene, 1-(2-bromoethyl)-4-fluorobenzene

020897-1G
FLUO020897-1GEA 36.9 EUR
FLUO020897-1G
4-Fluorphenethylbromid
4-Fluorphenethylbromid
Formel: C₈H₈BrF
Molecular Weight: 203,05 g/mol
MDL: MFCD03844794
CAS-Nummer: 332-42-3

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR