3',5'-Dihydroxyacetophenon

Lieferant: FLUOROCHEM

Synonyme: 1-(3,5-Dihydroxyphenyl)ethan-1-on

021304-25G
FLUO021304-25GEA 67.1 EUR
FLUO021304-25G
3',5'-Dihydroxyacetophenon
3',5'-Dihydroxyacetophenon
Formel: C₈H₈O₃
Molecular Weight: 152,15 g/mol
Siedepunkt: 296 °C (1013 hPa)
Schmelzpunkt: 148 °C
MDL: MFCD00002290
CAS-Nummer: 51863-60-6
EINECS: 257-480-1

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR