3,4-Dichlorthiobenzamid
Lieferant: FLUOROCHEM
Synonyme:
3,4-Dichlorthiobenzoesäureamid
021319-1G
FLUO021319-1GEA
107
EUR
FLUO021319-1G
3,4-Dichlorthiobenzamid
3,4-Dichlorthiobenzamid
Formel:
C₇H₅Cl₂NS |
MDL:
MFCD04973321 CAS-Nummer: 22179-73-3 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...