3,5-Dichlorthiobenzamid

Lieferant: FLUOROCHEM

Synonyme: 3,5-Dichlorthiobenzoesäureamid

021320-1G
FLUO021320-1GEA 142 EUR
FLUO021320-1G
3,5-Dichlorthiobenzamid
3,5-Dichlorthiobenzamid
Formel: C₇H₅Cl₂NS
Molecular Weight: 206.09 g/mol
MDL: MFCD04973322
CAS-Nummer: 22179-74-4

Bestellen

TECHNISCHE DATEN

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR