2,3-Dichlorthiobenzamid
Lieferant: FLUOROCHEM
Synonyme:
2,3-Dichlorthiobenzoesäureamid
021321-1G
FLUO021321-1GEA
130
EUR
FLUO021321-1G
2,3-Dichlorthiobenzamid
2,3-Dichlorthiobenzamid
Formel:
C₇H₅Cl₂NS Molecular Weight: 206,09 g/mol |
MDL:
MFCD00221163 CAS-Nummer: 84863-83-2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...