3-Chlorthiobenzamid
Lieferant: FLUOROCHEM
Synonyme:
3-Chlorthiobenzoesäureamid
021322-1G
FLUO021322-1GEA
0
EUR
FLUO021322-1G
3-Chlorthiobenzamid
3-Chlorthiobenzamid
Formel:
C₇H₆ClNS Molecular Weight: 171,65 g/mol |
MDL:
MFCD00276993 CAS-Nummer: 2548-79-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...