2-Fluorthiobenzamid
Lieferant: FLUOROCHEM
Synonyme:
2-Fluorthiobenzoesäureamid
021323-1G
FLUO021323-1GEA
107
EUR
FLUO021323-1G
2-Fluorthiobenzamid
2-Fluorthiobenzamid
Formel:
C₇H₆FNS Molecular Weight: 155.2 g/mol |
MDL:
MFCD04973316 CAS-Nummer: 75907-82-3 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...