4-Fluorthiobenzamid

Lieferant: FLUOROCHEM

Synonyme: 4-Fluorthiobenzoesäureamid

021324-1G 021324-5G
FLUO021324-1GEA 0 EUR
FLUO021324-1G FLUO021324-5G
4-Fluorthiobenzamid
4-Fluorthiobenzamid
Formel: C₇H₆FNS
Molecular Weight: 155.2 g/mol
MDL: MFCD01313285
CAS-Nummer: 22179-72-2
EINECS: 244-820-9
UN: 2811
ADR: 6.1 III

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR