3-Fluorthiobenzamid

Lieferant: FLUOROCHEM

Synonyme: 3-Fluorthiobenzoesäureamid

021325-1G
FLUO021325-1GEA 55.5 EUR
FLUO021325-1G
3-Fluorthiobenzamid
3-Fluorthiobenzamid
Formel: C₇H₆FNS
Molecular Weight: 155.2 g/mol
MDL: MFCD04973323
CAS-Nummer: 72505-20-5

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR