3-Methylthiobenzamid

Lieferant: FLUOROCHEM

Synonyme: Thio-m-toluamid, 3-Methylthiobenzoesäureamid

021340-1G
FLUO021340-1GEA 31.1 EUR
FLUO021340-1G
3-Methylthiobenzamid
3-Methylthiobenzamid
Formel: C₈H₉NS
Molecular Weight: 151,23 g/mol
MDL: MFCD01314035
CAS-Nummer: 2362-63-2

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR