2-Methylthiobenzamid

Lieferant: FLUOROCHEM

Synonyme: 2-Methylthiobenzoesäureamid, Thio-o-toluamid

021341-1G
FLUO021341-1GEA 39.9 EUR
FLUO021341-1G
2-Methylthiobenzamid
2-Methylthiobenzamid
Formel: C₈H₉NS
Molecular Weight: 151,23 g/mol
MDL: MFCD03945350
CAS-Nummer: 53515-19-8

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR