2-(Trifluormethyl)thiobenzamid
Lieferant: FLUOROCHEM
Synonyme:
2-(Trifluormethyl)thiobenzoesäureamid
021350-1G
FLUO021350-1GEA
91.2
EUR
FLUO021350-1G
2-(Trifluormethyl)thiobenzamid
2-(Trifluormethyl)thiobenzamid
Formel:
C₈H₆F₃NS Molecular Weight: 151,23 g/mol |
MDL:
MFCD04973326 CAS-Nummer: 886496-67-9 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...