2,3,4-Trifluorbenzolsulfonamid

Lieferant: FLUOROCHEM

Synonyme: 2,3,4-Trifluorbenzolsulfonsäureamid

021386-1G
FLUO021386-1GEA 116 EUR
FLUO021386-1G
2,3,4-Trifluorbenzolsulfonamid
2,3,4-Trifluorbenzolsulfonamid
Formel: C₆H₄F₃NO₂S
Molecular Weight: 211,16 g/mol
MDL: MFCD03094300
CAS-Nummer: 518070-13-8

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR