3-Chlor-2-fluorbenzolsulfonamid

Lieferant: FLUOROCHEM

021394-5G
FLUO021394-5GEA 200 EUR
FLUO021394-5G
3-Chlor-2-fluorbenzolsulfonamid
3-Chlor-2-fluorbenzolsulfonamid
Formel: C₆H₅ClFNO₂S
CAS-Nummer: 351003-58-2

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR