1-Brom-2-chlor-4-iodbenzol

Lieferant: FLUOROCHEM

021409-5G
FLUO021409-5GEA 48.8 EUR
FLUO021409-5G
1-Brom-2-chlor-4-iodbenzol
1-Bromo-2-chloro-4-iodobenzene
Formel: C₆H₃BrCl
Molecular Weight: 317,3500 g/mol
MDL: MFCD04973904
CAS-Nummer: 535934-25-9

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR