Ethyl-2-amino-4-methyl-3-thiophencarboxylat

Lieferant: FLUOROCHEM

Synonyme: Ethyl 2-amino-4-methylthiophene-3-carboxylate

021476-5G 021476-25G
FLUO021476-5GEA 96.5 EUR
FLUO021476-5G FLUO021476-25G
Ethyl-2-amino-4-methyl-3-thiophencarboxylat
Ethyl-2-amino-4-methyl-3-thiophencarboxylat
Formel: C₈H₁₁NO₂S
MDL: MFCD00051669
CAS-Nummer: 43088-42-2

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR