Ethyl-2-amino-4-phenyl-3-thiophencarboxylat
Lieferant: FLUOROCHEM
Synonyme:
Ethyl 2-Amino-4-phenylthiophene-3-carboxylate
021483-5G
FLUO021483-5GEA
180
EUR
FLUO021483-5G
Ethyl-2-amino-4-phenyl-3-thiophencarboxylat
Ethyl-2-amino-4-phenyl-3-thiophencarboxylat
Formel:
C₁₃H₁₃NO₂S Molecular Weight: 247,32 g/mol |
MDL:
MFCD00126391 CAS-Nummer: 4815-36-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...