Ethylpiperazin-1-carboxylat
Lieferant: FLUOROCHEM
Synonyme:
1-Ethoxycarbonyl piperazine, Ethyl N-piperazinecarboxylate
021571-100G
FLUO021571-100GEA
115
EUR
FLUO021571-100G
Ethylpiperazin-1-carboxylat
Ethylpiperazin-1-carboxylat
Formel:
C₇H₁₄N₂O₂ Molecular Weight: 158,2 g/mol |
MDL:
MFCD00005964 CAS-Nummer: 120-43-4 EINECS: 204-395-2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...