3,5-Dichlorphenylboronsäure

Lieferant: FLUOROCHEM
Achtung

021610-5G
FLUO021610-5GEA 42.9 EUR
FLUO021610-5G
3,5-Dichlorphenylboronsäure
3,5-Dichlorphenylboronsäure
Formel: C₆H₅BCl₂O₂
Molecular Weight: 190,82 g/mol
MDL: MFCD00051935
CAS-Nummer: 67492-50-6

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR