2,6-Dimethoxy-3-pyridinboronsäurepinakolester

Lieferant: FLUOROCHEM

021611-5G
FLUO021611-5GEA 944 EUR
FLUO021611-5G
2,6-Dimethoxy-3-pyridinboronsäurepinakolester
2,6-Dimethoxy-3-pyridinboronsäurepinakolester
Formel: C₁₃H₂₀BNO₄
MDL: MFCD03788240
CAS-Nummer: 214360-59-5

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR