4-(N,N-Dimethylamino)phenylboronsäurepinakolester

Lieferant: FLUOROCHEM

Synonyme: 4-(Dimethylamino)phenylboronic acid pinacol ester, N,N-Dimethyl-4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)aniline

021613-5G
FLUO021613-5GEA 282 EUR
FLUO021613-5G
4-(N,N-Dimethylamino)phenylboronsäurepinakolester
4-(N,N-Dimethylamino)phenylboronsäurepinakolester
Formel: C₁₄H₂₂BNO₂
Molecular Weight: 247,1410 g/mol
MDL: MFCD05663854
CAS-Nummer: 171364-78-6

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR