1-Isobutyl-1H-pyrazol-4-boronsäurepinakolester

Lieferant: FLUOROCHEM

Synonyme: 1-Isobutyl-4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)-1H-pyrazole, 1-Isobutylpyrazole-4-boronic acid, pinacol ester

021615-1G 021615-5G
FLUO021615-1GEA 138 EUR
FLUO021615-1G FLUO021615-5G
1-Isobutyl-1H-pyrazol-4-boronsäurepinakolester
1-Isobutyl-1H-pyrazol-4-boronsäurepinakolester
Formel: C₁₃H₂₃BN₂O₂
MDL: MFCD05663857
CAS-Nummer: 827614-66-4

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR