3,4-(Methylenedioxy)phenylboronsäurepinakolester
Lieferant: FLUOROCHEM
Synonyme:
5-(4,4,5,5-Tetramethyl-1,3,2-dioxaborolan-2-yl)-benzo-1,3-dioxole, 2-(Benzo[d][1,3]dioxol-5-yl)-4,4,5,5-tetramethyl-1,3,2-dioxaborolane, 3,4-Ethylenedioxybenzene boronic acid, pinacol ester
021632-5G
021632-10G
FLUO021632-5GEA
209
EUR
FLUO021632-5G
FLUO021632-10G
3,4-(Methylenedioxy)phenylboronsäurepinakolester
3,4-(Methylenedioxy)phenylboronsäurepinakolester
Formel:
C₁₃H₁₇BO₄ |
MDL:
MFCD05663889 CAS-Nummer: 94838-82-1 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...